Rink amide AM resin 200-400 mesh

Supplier: BACHEM AMERICAS INC

4014571.0001 4014571.0005 4014571.0025
D-1675.0001BAEA 211.77 CAD
D-1675.0001BA D-1675.0005BA D-1675.0025BA
Rink amide AM resin 200-400 mesh
Rink amide AM resin

Fmoc-Rink amide-AM resin, a standard resin for the Fmoc-SPPS of peptide amides. Cleavage was performed with 50 % TFA in CH₂Cl₂ or 95% aqueous TFA. Scavengers may be required. Bachem additionally offers the Rink amide (RAM) linker (Q-1660).


  • Substitution: 0.5-1.2 mmol/g

Storage Temperature: Refrigerator
MDL Number: MFCD00240214
CAS Number: 183599-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR