N-Fmoc-β-alanine 0,50-1,00 mmol/g on Wang resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine
4014393.0001
4014393.0005
D-1625.0001BAEA
185.26
CAD
D-1625.0001BA
D-1625.0005BA
N-Fmoc-β-alanine 0,50-1,00 mmol/g on Wang resin 200-400 mesh
N-Fmoc-β-alanine
Sequence: Fmoc-β-Ala-Wang resin (200-400 mesh)
CAS-No.:
Mol.weight:
SumFormula:
Bulk item no.: D-1625
Synonym(s): Fmoc-β-Ala-p-alkoxybenzyl alcohol resin
EINECS no.:
MDL no.: MFCD00801367
Substitution: 0.50-1.00 mmol/g
Sequence: Fmoc-β-Ala-Wang resin (200-400 mesh)
Substitution: 0.50-1.00 mmol/g
Formula:
C₁₈H₁₇NO₄ MW: 311.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063328 CAS Number: 35737-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...