Fmoc-Phe-Wang resin 0,5-1,0 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
4003165.0005
4003165.0050
D-1140.0005BAEA
335.58
CAD
D-1140.0005BA
D-1140.0050BA
Fmoc-Phe-Wang resin 0,5-1,0 mmol/g 200-400 mesh
Fmoc-Phe-Wang resin
Sequence: Fmoc-Phe-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Phe-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...