Fmoc-Ser(tBu) 0.5-1.0 mmol/g on Wang resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4006433.0001
4006433.0005
D-1070.0001BAEA
180.91
CAD
D-1070.0001BA
D-1070.0005BA
Fmoc-Ser(tBu) 0.5-1.0 mmol/g on Wang resin 200-400 mesh
Fmoc-Ser(tBu)
Sequence: Fmoc-Ser(tBu)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Ser(tBu)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...