Fmoc-Lys(boc)-Wang resin 0.5-1.0 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
4003241.0001
4003241.0005
D-1060.0001BAEA
180.91
CAD
D-1060.0001BA
D-1060.0005BA
Fmoc-Lys(boc)-Wang resin 0.5-1.0 mmol/g 200-400 mesh
Fmoc-Lys(boc)-Wang resin
Sequence: Fmoc-Lys(Boc)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Lys(Boc)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...