Fmoc-ε-aminocaproic acid-Wang resin 0.4-0.7 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
4006430.0001
4006430.0005
D-1050.0001BAEA
439.41
CAD
D-1050.0001BA
D-1050.0005BA
Fmoc-ε-aminocaproic acid-Wang resin 0.4-0.7 mmol/g 200-400 mesh
Fmoc-ε-aminocaproic acid-Wang resin
Sequence: Fmoc-ε-aminocaproic acid-Wang resin (200-400 mesh)
Synonym(s): Fmoc-6-aminohexanoic acid-Wang resin#Fmoc-6-aminohexanoic acid-p-alkoxybenzyl alcohol resin#Fmoc-εAhx-Wang resin#Fmoc-εAhx-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...