Z-D-allo-Thr(tBu)-OH·DCHA

Supplier: BACHEM AMERICAS INC

4044802.0001 4044802.0005
C-4210.0001BAEA 645.24 CAD
C-4210.0001BA C-4210.0005BA
Z-D-allo-Thr(tBu)-OH·DCHA
Z-D-allo-Thr(tBu)-OH·DCHA

Sequence: Z-D-allo-Thr(tBu)-OH · DCHA


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₂₈H₄₆N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD08458676
CAS Number: 100157-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR