Cbz-L-tyrosine benzyl ester

Supplier: BACHEM AMERICAS INC

4017301.0005 4017301.0025
C-4150.0005BAEA 722.85 CAD
C-4150.0005BA C-4150.0025BA
Cbz-L-tyrosine benzyl ester
Cbz-L-tyrosine benzyl ester

Sequence: Z-Tyr-OBzl


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₄H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00136670
CAS Number: 5513-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR