(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Lys(Fmoc)-OH, Cbz-Lys(Fmoc)-OH

4029328.0001 4029328.0005
C-4125.0001BAEA 309.81 CAD
C-4125.0001BA C-4125.0005BA
(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid
(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid

Sequence: Z-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065662
CAS Number: 105751-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR