Z-Asn-OMe

Supplier: BACHEM AMERICAS INC

4028527.0001 4028527.0005
C-4050.0001BAEA 180.91 CAD
C-4050.0001BA C-4050.0005BA
Z-Asn-OMe
Z-Asn-OMe

Sequence: Z-Asn-OMe


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₁₃H₁₆N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD02094371
CAS Number: 4668-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR