z-Tyr-OMe

Supplier: BACHEM AMERICAS INC

4000134.0025 4000134.0100
C-4040.0025BAEA 232.35 CAD
C-4040.0025BA C-4040.0100BA
z-Tyr-OMe
z-Tyr-OMe

Sequence: Z-Tyr-OMe


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₈H₁₉NO₅
Storage Temperature: Freezer
MDL Number: MFCD00038784
CAS Number: 13512-31-7
EINECS: 426-320-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR