z-trp(Boc)-OH·DCHA
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Trp(Boc)-OH·DCHA, Cbz-Trp(Boc)-OH·DCHA, Z-Trp(Boc)-OH.DCHA
4027713.0005
4027713.0025
C-3995.0005BAEA
439.41
CAD
C-3995.0005BA
C-3995.0025BA
z-trp(Boc)-OH·DCHA
z-trp(Boc)-OH·DCHA
Sequence: Z-Trp(Boc)-OH · DCHA
Formula:
C₃₆H₄₉N₃O₆ MW: 619.803 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01318754 CAS Number: 218938-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...