Z-Glu-OtBu

Supplier: BACHEM AMERICAS INC

4029292.0001 4029292.0005 4029292.0025
C-3905.0001BAEA 180.91 CAD
C-3905.0001BA C-3905.0005BA C-3905.0025BA
Z-Glu-OtBu
Z-Glu-OtBu

Sequence: Z-Glu-OtBu


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₇H₂₃NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077016
CAS Number: 5891-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR