Z-Asp(OMe)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (2S)-4-Methoxy-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid

4025293.0005 4025293.0025
C-3780.0005BAEA 413.58 CAD
C-3780.0005BA C-3780.0025BA
Z-Asp(OMe)-OH
Z-Asp(OMe)-OH

Sequence: Z-Asp(OMe)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₃H₁₅NO₆
MW: 281.264 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133585
CAS Number: 3160-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR