Z-Arg(Pbf)-OH · CHA

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Arg(Pbf)-OH.CHA, Cbz-Arg(Pbf)-OH·CHA

4019729.0001 4019729.0005
C-3715.0001BAEA 206.51 CAD
C-3715.0001BA C-3715.0005BA
Z-Arg(Pbf)-OH · CHA
Z-Arg(Pbf)-OH · CHA

Sequence: Z-Arg(Pbf)-OH · CHA


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₇H₃₆N₄O₇S·C₆H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00237327
CAS Number: 200190-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR