Z-Aib-OH

Supplier: BACHEM AMERICAS INC

4018563.0005 4018563.0025
C-3680.0005BAEA 309.81 CAD
C-3680.0005BA C-3680.0025BA
Z-Aib-OH
Z-Aib-OH

Sequence: Z-Aib-OH
Synonym(s): Z-α-Me-Ala-OH

Formula: C₁₂H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00004190
CAS Number: 15030-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR