(S)-2-(((Benzyloxy)carbonyl)amino)-4-((diphenyl(p-tolyl)methyl)amino)-4-oxobutanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Asn(Mtt)-OH

4017003.0005 4017003.0025
C-3630.0005BAEA 490.51 CAD
C-3630.0005BA C-3630.0025BA
(S)-2-(((Benzyloxy)carbonyl)amino)-4-((diphenyl(p-tolyl)methyl)amino)-4-oxobutanoic acid
(S)-2-(((Benzyloxy)carbonyl)amino)-4-((diphenyl(p-tolyl)methyl)amino)-4-oxobutanoic acid

Sequence: Z-Asn(Mtt)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₃₂H₃₀N₂O₅
MW: 522.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237170
CAS Number: 144317-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR