Z-Lys-NH₂·HCl

Supplier: BACHEM AMERICAS INC

4025068.0001 4025068.0005
C-3600.0001BAEA 180.91 CAD
C-3600.0001BA C-3600.0005BA
Z-Lys-NH₂·HCl
Z-Lys-NH₂·HCl

Sequence: Z-Lys-NH₂ · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₄H₂₂ClN₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00238483
CAS Number: 112785-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR