Z-Arg(Pmc)-OH · CHA

Supplier: BACHEM AMERICAS INC

4014138.0001 4014138.0005
C-3490.0001BAEA 206.51 CAD
C-3490.0001BA C-3490.0005BA
Z-Arg(Pmc)-OH · CHA
Z-Arg(Pmc)-OH · CHA

Sequence: Z-Arg(Pmc)-OH · CHA


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₈H₃₈N₄O₇S·C₆H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00153323
CAS Number: 112160-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR