1-Methyl-N-carbobenzoxy-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester, Z-Glu-OMe, 1-Methyl N-Cbz-L-glutamate, (S)-4-(((Benzyloxy)carbonyl)amino)-5-methoxy-5-oxopentanoic acid, N-Cbz-L-glutamic Acid 1-Methyl Ester

4016433.0005 4016433.0025
C-3465.0005BAEA 361.41 CAD
C-3465.0005BA C-3465.0025BA
1-Methyl-N-carbobenzoxy-L-glutamate
1-Methyl-N-carbobenzoxy-L-glutamate

Sequence: Z-Glu-OMe


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₄H₁₇NO₆
MW: 295.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083278
CAS Number: 5672-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR