Z-Lys(Z)-OSu

Supplier: BACHEM AMERICAS INC

4000136.0005 4000136.0025
C-2930.0005BAEA 180.91 CAD
C-2930.0005BA C-2930.0025BA
Z-Lys(Z)-OSu
Z-Lys(Z)-OSu

Sequence: Z-Lys(Z)-OSu


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₆H₂₉N₃O₈
MW: 511.53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00057964
CAS Number: 21160-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR