Z-Lys(Z)-OH
Supplier: BACHEM AMERICAS INC
4000512.0025
4000512.0100
C-2925.0025BAEA
206.51
CAD
C-2925.0025BA
C-2925.0100BA
Z-Lys(Z)-OH
Z-Lys(Z)-OH
Sequence: Z-Lys(Z)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₂₂H₂₆N₂O₆ MW: 414.458 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00037822 CAS Number: 405-39-0 EINECS: 206-971-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...