Z-Val-Ala-OH

Supplier: BACHEM AMERICAS INC

4007093.0005 4007093.0025
C-2835.0005BAEA 309.81 CAD
C-2835.0005BA C-2835.0025BA
Z-Val-Ala-OH
Proteins and Peptides

Sequence: Z-Val-Ala-OH

Formula: C₁₆H₂₂N₂O₅
MDL Number: MFCD00020395
CAS Number: 24787-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR