N-Carbobenzyloxy-L-valine

Supplier: BACHEM AMERICAS INC

4000403.0025 4000403.0100
C-2805.0025BAEA 175.66 CAD
C-2805.0025BA C-2805.0100BA
N-Carbobenzyloxy-L-valine
N-Carbobenzyloxy-L-valine

Sequence: Z-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₁₃H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00008922
CAS Number: 1149-26-4
EINECS: 214-562-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR