Z-Tyr-Ala-OH

Supplier: BACHEM AMERICAS INC

4014005.0001 4014005.0005
C-2750.0001BAEA 309.81 CAD
C-2750.0001BA C-2750.0005BA
Z-Tyr-Ala-OH
Z-Tyr-Ala-OH

Sequence: Z-Tyr-Ala-OH

Formula: C₂₀H₂₂N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00037236
CAS Number: 23018-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR