N-Carbobenzoxy-L-tyrosine

Supplier: BACHEM AMERICAS INC

4001149.0025 4001149.0100
C-2730.0025BAEA 211.78 CAD
C-2730.0025BA C-2730.0100BA
N-Carbobenzoxy-L-tyrosine
N-Carbobenzoxy-L-tyrosine

Sequence: Z-Tyr-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₇H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD00037180
CAS Number: 1164-16-5
EINECS: 214-609-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR