Z-Trp-OSu

Supplier: BACHEM AMERICAS INC

4008236.0005 4008236.0025
C-2680.0005BAEA 175.66 CAD
C-2680.0005BA C-2680.0025BA
Z-Trp-OSu
Z-Trp-OSu

Sequence: Z-Trp-OSu

Formula: C₂₃H₂₁N₃O₆
Storage Temperature: Freezer
MDL Number: MFCD00153347
CAS Number: 50305-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR