N(α)-Cbz-D-tryptophan

Supplier: BACHEM AMERICAS INC

4001610.0005 4001610.0025
C-2670.0005BAEA 227.28 CAD
C-2670.0005BA C-2670.0025BA
N(α)-Cbz-D-tryptophan
N(α)-Cbz-D-tryptophan

Sequence: Z-D-Trp-OH

Formula: C₁₉H₁₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00037945
CAS Number: 2279-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR