N(α)-Cbz-L-tryptophan
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid, N(α)-Benzyloxycarbonyl-L-tryptophan, N-Cbz-L-Tryptophan, Z-Trp-OH
4001218.0025
4001218.0100
C-2665.0025BAEA
175.66
CAD
C-2665.0025BA
C-2665.0100BA
N(α)-Cbz-L-tryptophan
N(α)-Cbz-L-tryptophan
Sequence: Z-Trp-OH
Formula:
C₁₉H₁₈N₂O₄ MW: 338.4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065700 CAS Number: 7432-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...