N-Carbobenzoxy-L-serine benzyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl, Benzyl ((benzyloxy)carbonyl)-L-serinate

4004231.0005 4004231.0025
C-2600.0005BAEA 206.51 CAD
C-2600.0005BA C-2600.0025BA
N-Carbobenzoxy-L-serine benzyl ester
N-Carbobenzoxy-L-serine benzyl ester

Sequence: Z-Ser-OBzl


Bachem also offers derivatives of phosphoserine and sulfoserine.


Educt for the synthesis of racemic Gla derivatives. The versatile intermediate Z-Dha-OBzl can be obtained by treating Z-Ser-OBzl with DCC and CuCl.

Formula: C₁₈H₁₉NO₅
MW: 329.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037824
CAS Number: 21209-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR