N-Cbz-L-serine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

4001465.0025 4001465.0100
C-2590.0025BAEA 175.66 CAD
C-2590.0025BA C-2590.0100BA
N-Cbz-L-serine
N-Cbz-L-serine

Sequence: Z-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


Educt in Hoveyda's synthesis of L-3-fluoroalanine and N-methyl-L-3-fluoroalanine derivatives via the oxazolidinone.

Formula: C₁₁H₁₃NO₅
MW: 239.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002662
CAS Number: 1145-80-8
EINECS: 214-546-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR