2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid

Supplier: BACHEM AMERICAS INC

4001068.0025 4001068.0100
C-2570.0025BAEA 439.41 CAD
C-2570.0025BA C-2570.0100BA
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid

Sequence: Z-Sar-OH


Bachem also offers Fmoc-Sar-Wang resin (D-1805) and Fmoc-Sar-SASRIN resin (-D-1645).


Synonym(s): Z-N-Me-Gly-OH

Formula: C₁₁H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00021747
CAS Number: 39608-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR