Z-Pyr-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-1-Carbobenzoxy-5-oxopyrrolidine-2-carboxylic acid, N-Cbz-L-pyroglutamic acid, (S)-1-Benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, Z-Pyr-OH, N-Benzyloxycarbonyl-L-pyroglutamic acid, N-Carbobenzoxy-L-pyroglutamic acid, (S)-1-Cbz-5-oxopyrrolidine-2-carboxylic acid, (S)-1-((Benzyloxy)carbonyl)-5-oxopyrrolidine-2-carboxylic acid
Sequence: Z-Pyr-OH
Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.
Formula:
C₁₃H₁₃NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00037352 CAS Number: 32159-21-0 EINECS: 250-936-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...