Z-Pro-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-1-((S)-1-((Benzyloxy)carbonyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxylic acid

4000216.0005 4000216.0025
C-2545.0005BAEA 309.81 CAD
C-2545.0005BA C-2545.0025BA
Z-Pro-Pro-OH
Z-Pro-Pro-OH

Sequence: Z-Pro-Pro-OH

Formula: C₁₈H₂₂N₂O₅
MW: 346.383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077035
CAS Number: 7360-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR