Z-Pro-OSu

Supplier: BACHEM AMERICAS INC

4000215.0005 4000215.0025
C-2475.0005BAEA 175.66 CAD
C-2475.0005BA C-2475.0025BA
Z-Pro-OSu
Z-Pro-OSu

Sequence: Z-Pro-OSu

Formula: C₁₇H₁₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00042757
CAS Number: 3397-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR