N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine
Supplier: BACHEM AMERICAS INC
4000601.0005
4000601.0025
C-2420.0005BAEA
313.55
CAD
C-2420.0005BA
C-2420.0025BA
N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine
N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine
Sequence: Z-Phe-Phe-OH
Substrate for cathepsin A. As Fmoc-Phe-Phe-OH (B-2150), Z-Phe-Phe-OH forms hydrogels
Formula:
C₂₆H₂₆N₂O₅ Storage Temperature: Freezer |
MDL Number:
MFCD00191139 CAS Number: 13122-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...