Z-Phe-ala

Supplier: BACHEM AMERICAS INC

4000227.0005 4000227.0025
C-2360.0005BAEA 325.59 CAD
C-2360.0005BA C-2360.0025BA
Z-Phe-ala

Sequence: Z-Phe-Ala-OH


Sensitive substrate for cathepsin A.

Formula: C₂₀H₂₂N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00037237
CAS Number: 21881-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR