(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH
4000222.0025
4000222.0100
C-2310.0025BAEA
175.62
CAD
C-2310.0025BA
C-2310.0100BA
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Sequence: Z-Phe-OH
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Metalloendoprotease inhibitor.
Formula:
C₁₇H₁₇NO₄ MW: 299.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00020418 CAS Number: 1161-13-3 EINECS: 214-599-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...