Cbz-L-ornithine

Supplier: BACHEM AMERICAS INC

4000190.0005 4000190.0025
C-2305.0005BAEA 361.41 CAD
C-2305.0005BA C-2305.0025BA
Cbz-L-ornithine
Cbz-L-ornithine

Sequence: Z-Orn-OH

Formula: C₁₃H₁₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00038200
CAS Number: 2640-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR