N-Cbz-N-methyl-L-leucine

Supplier: BACHEM AMERICAS INC

Synonyms: Z-N-Me-Leu-OH

4010008.0001 4010008.0005
C-2240.0001BAEA 180.91 CAD
C-2240.0001BA C-2240.0005BA
N-Cbz-N-methyl-L-leucine
N-Cbz-N-methyl-L-leucine

Sequence: Z-N-Me-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038521
CAS Number: 33099-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR