N(α)-Benzyloxycarbonyl-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH

4001304.0005 4001304.0025
C-2200.0005BAEA 180.14 CAD
C-2200.0005BA C-2200.0025BA
N(α)-Benzyloxycarbonyl-L-lysine
N(α)-Benzyloxycarbonyl-L-lysine

Sequence: Z-Lys-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038204
CAS Number: 2212-75-1
EINECS: 218-662-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR