N-Carbobenzoxy-L-leucine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Cbz-L-leucine, Z-Leu-OH
4000522.0025
4000522.0100
C-2100.0025BAEA
175.66
CAD
C-2100.0025BA
C-2100.0100BA
N-Carbobenzoxy-L-leucine
N-Carbobenzoxy-L-leucine
Sequence: Z-Leu-OH (oil)
Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00026494 CAS Number: 2018-66-8 EINECS: 217-960-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...