N(α)-Cbz-L-histidine
Supplier: BACHEM AMERICAS INC
Synonyms:
Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH
4000625.0005
4000625.0025
C-1945.0005BAEA
175.66
CAD
C-1945.0005BA
C-1945.0025BA
N(α)-Cbz-L-histidine
N(α)-Cbz-L-histidine
Sequence: Z-His-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Formula:
C₁₄H₁₅N₃O₄ MW: 289.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065960 CAS Number: 14997-58-1 EINECS: 239-084-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...