Z-Gly-Pro

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-1-(2-(((Benzyloxy)carbonyl)amino)acetyl)pyrrolidine-2-carboxylic acid, Z-Gly-Pro-OH

4000301.0005 4000301.0025
C-1880.0005BAEA 206.51 CAD
C-1880.0005BA C-1880.0025BA
Z-Gly-Pro
Z-Gly-Pro

Sequence: Z-Gly-Pro-OH


Cleavage product of the prolyl endopeptidase substrate M-1265.

Formula: C₁₅H₁₈N₂O₅
MW: 306.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037341
CAS Number: 1160-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR