N-[(Benzyloxy)carbonyl]glycylglycylphenylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Gly-Gly-Phe-OH
4010787.0005
4010787.0025
C-1795.0005BAEA
2098.54
CAD
C-1795.0005BA
C-1795.0025BA
N-[(Benzyloxy)carbonyl]glycylglycylphenylalanine
N-[(Benzyloxy)carbonyl]glycylglycylphenylalanine
Sequence: Z-Gly-Gly-Phe-OH
Formula:
C₂₁H₂₃N₃O₆ MW: 413.43 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00055796 CAS Number: 13171-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...