Z-Gly-Gly-Gly-Gly-Gly-OH

Supplier: BACHEM AMERICAS INC

4006909.0001 4006909.0005
C-1755.0001BAEA 464.74 CAD
C-1755.0001BA C-1755.0005BA
Z-Gly-Gly-Gly-Gly-Gly-OH
Z-Gly-Gly-Gly-Gly-Gly-OH

Sequence: Z-Gly-Gly-Gly-Gly-Gly-OH


Z-pentaglycine, Z-GGGGG.

Formula: C₁₈H₂₃N₅O₈
MW: 437.409 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077019
CAS Number: 20228-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR