Z-Gly-Gly-OSu

Supplier: BACHEM AMERICAS INC

4000154.0005 4000154.0025
C-1710.0005BAEA 413.58 CAD
C-1710.0005BA C-1710.0025BA
Z-Gly-Gly-OSu
Z-Gly-Gly-OSu

Sequence: Z-Gly-Gly-OSu

Formula: C₁₆H₁₇N₃O₇
MW: 363.326 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00191098
CAS Number: 32943-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR