(Carbobenzoxy)glycylglycine

Supplier: BACHEM AMERICAS INC

Synonyms: 2-(2-(((Benzyloxy)carbonyl)amino)acetamido)acetic acid, Z-Gly-Gly-OH, (Carbobenzoxy)glycylglycine

4000488.0025 4000488.0100
C-1700.0025BAEA 614.51 CAD
C-1700.0025BA C-1700.0100BA
(Carbobenzoxy)glycylglycine
(Carbobenzoxy)glycylglycine

Sequence: Z-Gly-Gly-OH

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037783
CAS Number: 2566-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR