Z-Gly-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Gly-Osu

4000533.0025 4000533.0100
C-1665.0025BAEA 206.51 CAD
C-1665.0025BA C-1665.0100BA
Z-Gly-OSu
Z-Gly-OSu

Sequence: Z-Gly-OSu


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₁₄H₁₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00153332
CAS Number: 2899-60-7
EINECS: 220-789-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR