N-Cbz-glycine(Z-Gly-OH)

Supplier: BACHEM AMERICAS INC

Synonyms: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

4000228.0100 4000228.0500
C-1655.0100BAEA 201.47 CAD
C-1655.0100BA C-1655.0500BA
N-Cbz-glycine(Z-Gly-OH)
N-Cbz-Glycine (Z-Gly-OH)

Sequence: Z-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002691
CAS Number: 1138-80-3
EINECS: 214-516-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR